siRNA Details
| Gene Symbol: PRICKLE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144881.1 | NP_001138353.1 | 3, 4 | 541 | ||
| NM_001144882.1 | NP_001138354.1 | 3, 4 | 540 | |||
| NM_001144883.1 | NP_001138355.1 | 3, 4 | 537 | |||
| NM_153026.2 | NP_694571.2 | 3, 4 | 608 | |||
| XM_011537946.1 | XP_011536248.1 | 3, 4 | 359 | |||
| XM_011537947.2 | XP_011536249.1 | 3, 4 | 590 | |||
| XM_017018838.1 | XP_016874327.1 | 3, 4 | 404 | |||
| XM_017018839.1 | XP_016874328.1 | 3, 4 | 604 | |||
| XM_017018840.1 | XP_016874329.1 | 3, 4 | 424 | |||
| GenBank mRNA | AF399844.1 | - | 3, 4 | 514 | ||
| AF543759.1 | - | 3, 4 | 381 | |||
| AF543760.1 | - | 4, 5 | 635 | |||
| AK056499.1 | - | 3, 4 | 514 | |||
| AK122877.1 | - | 3, 4 | 542 | |||
| AK122881.1 | - | 3, 4 | 485 | |||
| BC114939.1 | - | 3, 4 | 449 | 40084889 | ||
| BC114940.1 | - | 3, 4 | 450 | 40084893 | ||
| CN390789.1 | - | 2, 3 | 201 | |||
| DA321918.1 | - | 3, 4 | 540 | |||

PRICKLE1
Genomic Map