siRNA Details
| Gene Symbol: AMZ2P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026903.1 | 1 | 553 | |||
| GenBank mRNA | AK056627.1 | - | 1 | 536 | ||
| BC073870.1 | - | 1 | 18 | |||
| BX537538.1 | - | 1 | 427 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AMZ2P1| Gene Symbol: AMZ2P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026903.1 | 1 | 553 | |||
| GenBank mRNA | AK056627.1 | - | 1 | 536 | ||
| BC073870.1 | - | 1 | 18 | |||
| BX537538.1 | - | 1 | 427 | |||