siRNA Details
| Gene Symbol: TMEM268 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153045.3 | NP_694590.2 | 9 | 1303 | ||
| XM_006716997.1 | XP_006717060.1 | 9 | 1306 | |||
| XM_011518350.1 | XP_011516652.1 | 10 | 1474 | |||
| XM_011518351.2 | XP_011516653.1 | 10 | 1602 | |||
| XM_011518352.1 | XP_011516654.1 | 10 | 1471 | |||
| XM_011518353.1 | XP_011516655.1 | 9 | 1363 | |||
| XM_011518354.1 | XP_011516656.1 | 9 | 1509 | |||
| XM_011518355.1 | XP_011516657.1 | 9 | 1506 | |||
| XM_017014429.1 | XP_016869918.1 | 10 | 1630 | |||
| XM_017014430.1 | XP_016869919.1 | 8 | 1395 | |||
| GenBank mRNA | AK095364.1 | - | 4 | 627 | ||
| AK289383.1 | - | 9 | 1272 | |||
| AL834412.1 | - | 7 | 915 | |||
| BC019913.1 | - | 3 | 476 | |||
| BC062550.1 | - | 5 | 645 | |||
| BC107134.1 | - | 9 | 1454 | 40025600 | ||
| BX649023.1 | - | 9 | 1189 | |||

TMEM268
Genomic Map