siRNA Details
| Gene Symbol: CCR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164680.1 | NP_001158152.1 | 3 | 425 | ||
| NM_001837.3 | NP_001828.1 | 3 | 444 | |||
| NM_178328.1 | NP_847898.1 | 3 | 434 | |||
| NM_178329.2 | NP_847899.1 | 2 | 365 | |||
| XM_006712960.3 | XP_006713023.1 | 3 | 471 | |||
| XM_011533335.2 | XP_011531637.1 | 3 | 551 | |||
| XM_017005685.1 | XP_016861174.1 | 3 | 1354 | |||
| XM_017005686.1 | XP_016861175.1 | 4 | 1535 | |||
| GenBank mRNA | AF026535.1 | - | 1 | 196 | ||
| AF262299.1 | - | 2 | 315 | |||
| AF262300.1 | - | 3 | 1299 | |||
| AF262301.1 | - | 3 | 355 | |||
| AF262302.1 | - | 3 | 365 | |||
| BC110297.1 | - | 2 | 274 | 5212541 | ||
| BC130318.1 | - | 2 | 276 | 40146349 | ||
| BC130320.1 | - | 2 | 276 | 40146351 | ||
| U28694.1 | - | 1 | 227 | |||

CCR3
Genomic Map