siRNA Details
| Gene Symbol: MFSD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152778.2 | NP_689991.1 | 8 | 892 | ||
| XM_005262893.1 | XP_005262950.1 | 7 | 806 | |||
| XM_005262896.1 | XP_005262953.1 | 6 | 659 | |||
| XM_005262898.2 | XP_005262955.1 | 7 | 806 | |||
| XM_011531830.1 | XP_011530132.1 | 6 | 692 | |||
| XM_011531832.1 | XP_011530134.1 | 6 | 692 | |||
| GenBank mRNA | AK074564.1 | - | 7 | 741 | ||
| AK294439.1 | - | 6 | 624 | |||
| AK294527.1 | - | 6 | 570 | |||
| AK294957.1 | - | 7 | 734 | |||
| AK315596.1 | - | 7 | 738 | |||
| BC029503.1 | - | 8 | 857 | 5272664 | ||

MFSD8
Genomic Map