siRNA Details
| Gene Symbol: PRSS33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152891.2 | NP_690851.2 | 6 | 1550 | ||
| XM_011522451.2 | XP_011520753.1 | 7 | 1674 | |||
| XM_011522452.2 | XP_011520754.1 | 7 | 1486 | |||
| GenBank mRNA | AF536382.1 | - | 6 | 1457 | ||
| BC036846.1 | - | 7 | 1532 | 5245847 | ||
| BC062334.1 | - | 6 | 1552 | 6110198 | ||

PRSS33
Genomic Map