siRNA Details
| Gene Symbol: LRWD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317721.1 | NP_001304650.1 | 3 | 599 | ||
| NM_152892.2 | NP_690852.1 | 3 | 581 | |||
| XM_017011845.1 | XP_016867334.1 | 3 | 482 | |||
| GenBank mRNA | AB209379.1 | - | 3 | 459 | ||
| AK290967.1 | - | 3 | 480 | |||
| AK313771.1 | - | 3 | 464 | |||
| AL133057.1 | - | 3 | 531 | |||
| BC009436.1 | - | 2 | 272 | |||
| BC018769.1 | - | 2 | 111 | |||
| BC030547.1 | - | 3 | 521 | 5170588 | ||

LRWD1
Genomic Map