siRNA Details
| Gene Symbol: ZNF548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172773.1 | NP_001166244.1 | 4 | 1746 | ||
| NM_152909.3 | NP_690873.2 | 3 | 1710 | |||
| XM_017026316.1 | XP_016881805.1 | 6 | 2016 | |||
| GenBank mRNA | AK057494.1 | - | 4 | 1699 | ||
| AK299069.1 | - | 4 | 1593 | |||
| BC030788.2 | - | 3 | 1710 | 5266647 | ||

ZNF548
Genomic Map