siRNA Details
| Gene Symbol: TMEM130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134450.1 | NP_001127922.1 | 8 | 2288 | ||
| NM_001134451.1 | NP_001127923.1 | 7 | 1946 | |||
| NM_152913.2 | NP_690877.1 | 8 | 2252 | |||
| GenBank mRNA | AB072903.1 | - | 3 | 1094 | ||
| AK124634.1 | - | 8 | 3119 | |||
| AK225783.1 | - | 8 | 2221 | |||
| AL832523.1 | - | 7 | 1976 | |||
| AL834348.1 | - | 8 | 2088 | |||
| AL834349.1 | - | 8 | 2102 | |||
| AY358583.1 | - | 9 | 2174 | |||
| BC030793.1 | - | 8 | 2211 | 5263130 | ||
| BC037895.1 | - | 7 | 1846 | 5275566 | ||

TMEM130
Genomic Map