siRNA Details
| Gene Symbol: CCDC140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153038.1 | NP_694583.1 | 2 | 1132 | ||
| XM_017003466.1 | XP_016858955.1 | 2 | 973 | |||
| GenBank mRNA | AK057009.1 | - | 2 | 1132 | ||
| BC109388.1 | - | 2 | 1037 | 40023803 | ||

CCDC140
Genomic Map