siRNA Details
| Gene Symbol: TMEM92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168215.1 | NP_001161687.1 | 4 | 227 | ||
| NM_153229.2 | NP_694961.2 | 3 | 192 | |||
| XM_006721730.3 | XP_006721793.1 | 3 | 192 | |||
| GenBank mRNA | AK090637.1 | - | 3 | 147 | ||
| AY358204.1 | - | 4 | 227 | |||
| BC064507.1 | - | 3 | 120 | 6455254 | ||
| DB072318.1 | - | 3 | 193 | |||

TMEM92
Genomic Map