siRNA Details
| Gene Symbol: LRRC57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153260.2 | NP_694992.2 | 3 | 264 | ||
| XM_011521423.2 | XP_011519725.1 | 2 | 152 | |||
| XM_011521424.2 | XP_011519726.1 | 2 | 152 | |||
| GenBank mRNA | AK094131.1 | - | 3 | 258 | ||
| AK094891.1 | - | 2 | 498 | |||
| AK309861.1 | - | 3 | 253 | |||
| BC058935.1 | - | 3 | 220 | 5578024 | ||
| BX538324.1 | - | 2 | 410 | |||
| CR749223.1 | - | 3 | 233 | |||
| DA749946.1 | - | 3 | 264 | |||

LRRC57
Genomic Map