siRNA Details
| Gene Symbol: CNEP1R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281789.1 | NP_001268718.1 | 3, 4 | 289 | ||
| NM_153261.5 | NP_694993.2 | 4, 5 | 340 | |||
| GenBank mRNA | AK095420.1 | - | 3, 4 | 262 | ||
| AK301610.1 | - | 4, 5 | 305 | |||
| AK307132.1 | - | 3, 4 | 336 | |||
| BC036683.2 | - | 4, 5 | 269 | 5266084 | ||
| CR933600.1 | - | 4, 5 | 285 | |||
| FJ516381.1 | - | 3, 4 | 217 | |||
| HY011229.1 | - | 3, 4 | 290 | |||

CNEP1R1
Genomic Map