siRNA Details
| Gene Symbol: TMEM151A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153266.3 | NP_694998.1 | 2 | 576 | ||
| GenBank mRNA | AL834459.1 | - | 1 | 374 | ||
| BC033898.2 | - | 2 | 607 | 4813116 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM151A| Gene Symbol: TMEM151A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153266.3 | NP_694998.1 | 2 | 576 | ||
| GenBank mRNA | AL834459.1 | - | 1 | 374 | ||
| BC033898.2 | - | 2 | 607 | 4813116 | ||