siRNA Details
| Gene Symbol: MAMDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153267.4 | NP_694999.3 | 11 | 2185 | ||
| NR_125850.1 | 11 | 2185 | ||||
| XM_011518471.1 | XP_011516773.1 | 11 | 2247 | |||
| XM_011518475.2 | XP_011516777.1 | 11 | 2247 | |||
| XM_017014587.1 | XP_016870076.1 | 11 | 2247 | |||
| GenBank mRNA | AK304040.1 | - | 11 | 1945 | ||
| AK304531.1 | - | 11 | 1773 | |||
| BC015417.2 | - | 3 | 249 | |||
| BC016383.1 | - | 11 | 1903 | 4696722 | ||
| BC063634.1 | - | 11 | 2162 | 4399866 | ||

MAMDC2
Genomic Map