siRNA Details
Gene Symbol: KLHL34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153270.2 | NP_695002.1 | 1 | 2119 | ||
GenBank mRNA | AK092279.1 | - | 1 | 2062 | ||
BC109031.2 | - | 1 | 2031 | 40027951 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KLHL34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153270.2 | NP_695002.1 | 1 | 2119 | ||
GenBank mRNA | AK092279.1 | - | 1 | 2062 | ||
BC109031.2 | - | 1 | 2031 | 40027951 |