siRNA Details
| Gene Symbol: PSTK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153336.2 | NP_699167.2 | 1 | 536 | ||
| XM_017015640.1 | XP_016871129.1 | 1 | 162 | |||
| XM_017015641.1 | XP_016871130.1 | 1 | 162 | |||
| GenBank mRNA | AK127173.1 | - | 1 | 162 | ||
| BC035344.1 | - | 1 | 536 | 5185692 | ||

PSTK
Genomic Map