siRNA Details
Gene Symbol: ZNF791 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153358.2 | NP_699189.2 | 3, 4 | 348 | ||
XM_017026412.1 | XP_016881901.1 | 3, 4 | 318 | |||
GenBank mRNA | AK074877.1 | - | 3, 4 | 327 | ||
AK297715.1 | - | 2, 3 | 185 | |||
AK298597.1 | - | 4, 5 | 486 | |||
BC106937.1 | - | 3, 4 | 282 | 40033817 | ||
BC106938.1 | - | 3, 4 | 281 | 40033819 | ||
DC329382.1 | - | 3, 4 | 348 |
