siRNA Details
| Gene Symbol: LIX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153234.4 | NP_694966.3 | 2 | 383 | ||
| GenBank mRNA | AK098400.1 | - | 2 | 439 | ||
| AK291034.1 | - | 2 | 366 | |||
| AK307885.1 | - | 2 | 324 | |||
| BC036467.2 | - | 2 | 302 | 5264055 | ||

LIX1
Genomic Map