siRNA Details
| Gene Symbol: C6orf223 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171992.2 | NP_001165463.1 | 4 | 1003 | ||
| NM_001324369.1 | NP_001311298.1 | 3 | 833 | |||
| NM_153246.5 | NP_694978.2 | 4 | 960 | |||
| XM_017010425.1 | XP_016865914.1 | 4 | 2042 | |||
| GenBank mRNA | AL555526.3 | - | 3 | 801 | ||
| BC029071.2 | - | 4 | 970 | 4341806 | ||
| BC032706.1 | - | 3 | 1274 | 5497384 | ||

C6orf223
Genomic Map