siRNA Details
| Gene Symbol: CYBB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000397.3 | NP_000388.2 | 5 | 509 | ||
| GenBank mRNA | AK289753.1 | - | 5 | 494 | ||
| AK301491.1 | - | 4 | 371 | |||
| AK312929.1 | - | 5 | 469 | |||
| BC032720.1 | - | 5 | 495 | 5505170 | ||
| DA995835.1 | - | 5 | 509 | |||
| KU178009.1 | - | 5 | 448 | |||
| KU178010.1 | - | 5 | 448 | |||
| X04011.1 | - | 5 | 463 | |||

CYBB
Genomic Map