siRNA Details
| Gene Symbol: ZNF549 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199295.1 | NP_001186224.1 | 4 | 716 | ||
| NM_153263.2 | NP_694995.2 | 3 | 677 | |||
| XM_006723128.3 | XP_006723191.1 | 4 | 791 | |||
| GenBank mRNA | AK092236.1 | - | 3 | 677 | ||
| AK122749.1 | - | 4 | 650 | |||
| BC060863.1 | - | 4 | 620 | 30340439 | ||

ZNF549
Genomic Map