siRNA Details
| Gene Symbol: PSTK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153336.2 | NP_699167.2 | 4 | 1213 | ||
| XM_017015640.1 | XP_016871129.1 | 4 | 839 | |||
| GenBank mRNA | AK094271.1 | - | 3 | 1318 | ||
| AK127173.1 | - | 4 | 839 | |||
| BC035344.1 | - | 4 | 1213 | 5185692 | ||

PSTK
Genomic Map