siRNA Details
| Gene Symbol: LRRC34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172779.1 | NP_001166250.1 | 3 | 571 | ||
| NM_001172780.1 | NP_001166251.1 | 3 | 571 | |||
| NM_153353.4 | NP_699184.2 | 3 | 571 | |||
| XM_011512442.2 | XP_011510744.1 | 3 | 355 | |||
| XM_017005746.1 | XP_016861235.1 | 3 | 529 | |||
| GenBank mRNA | AK095125.1 | - | 3 | 316 | ||
| AK293661.1 | - | 3 | 449 | |||
| AK295065.1 | - | 3 | 310 | |||
| AK302466.1 | - | 3 | 333 | |||
| BC032841.2 | - | 3 | 310 | 4826965 | ||

LRRC34
Genomic Map