siRNA Details
| Gene Symbol: ZNF679 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153363.2 | NP_699194.2 | 5 | 692 | ||
| XM_017011797.1 | XP_016867286.1 | 5 | 653 | |||
| GenBank mRNA | BC033523.1 | - | 5 | 693 | 4821935 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF679| Gene Symbol: ZNF679 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153363.2 | NP_699194.2 | 5 | 692 | ||
| XM_017011797.1 | XP_016867286.1 | 5 | 653 | |||
| GenBank mRNA | BC033523.1 | - | 5 | 693 | 4821935 | |