siRNA Details
| Gene Symbol: LACC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128303.1 | NP_001121775.1 | 7 | 2313 | ||
| NM_153218.2 | NP_694950.2 | 7 | 2149 | |||
| XM_005266261.3 | XP_005266318.1 | 7 | 2772 | |||
| XM_006719766.3 | XP_006719829.1 | 7 | 1930 | |||
| XM_006719767.3 | XP_006719830.1 | 6 | 1988 | |||
| XM_011534933.2 | XP_011533235.1 | 8 | 2967 | |||
| XM_011534934.2 | XP_011533236.1 | 7 | 2086 | |||
| XM_017020394.1 | XP_016875883.1 | 7 | 2206 | |||
| GenBank mRNA | AK096044.1 | - | 7 | 2149 | ||
| BX538036.1 | - | 3 | 884 | |||

LACC1
Genomic Map