siRNA Details
| Gene Symbol: MCOLN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153259.3 | NP_694991.2 | 14 | 2338 | ||
| XM_005270719.3 | XP_005270776.1 | 14 | 2115 | |||
| XM_006710552.2 | XP_006710615.1 | 15 | 2428 | |||
| XM_011541187.2 | XP_011539489.1 | 14 | 2172 | |||
| XM_011541188.2 | XP_011539490.1 | 14 | 2455 | |||
| GenBank mRNA | AK094010.1 | - | 12 | 2013 | ||
| AK129786.1 | - | 8 | 1341 | |||
| AV713773.2 | - | 1 | 40 | |||

MCOLN2
Genomic Map