siRNA Details
| Gene Symbol: SNX33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318146.1 | NP_001305075.1 | 1 | 836 | ||
| NM_153271.1 | NP_695003.1 | 1 | 836 | |||
| GenBank mRNA | AK091291.1 | - | 1 | 88 | ||
| AL833039.1 | - | 1 | 800 | |||
| BC018775.1 | - | 1 | 836 | 4869639 | ||
| EF219141.1 | - | 1 | 761 | |||
| EF653821.1 | - | 1 | 1002 | |||

SNX33
Genomic Map