siRNA Details
| Gene Symbol: BEND2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184767.1 | NP_001171696.1 | 9, 10 | 1627 | ||
| NM_153346.4 | NP_699177.2 | 11, 12 | 1900 | |||
| XM_011545447.1 | XP_011543749.1 | 11, 12 | 1924 | |||
| XM_017029266.1 | XP_016884755.1 | 10, 11 | 1798 | |||
| XM_017029267.1 | XP_016884756.1 | 11, 12 | 1924 | |||
| GenBank mRNA | AK128155.1 | - | 11, 12 | 1882 | ||
| BC037301.2 | - | 11, 12 | 1880 | 4827886 | ||
| BC096745.1 | - | 9, 10 | 1560 | 4830576 | ||

BEND2
Genomic Map