siRNA Details
| Gene Symbol: FBXW8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012174.1 | NP_036306.1 | 5, 6 | 723 | ||
| NM_153348.2 | NP_699179.2 | 5, 6 | 921 | |||
| XM_005253867.2 | XP_005253924.1 | 4, 5 | 779 | |||
| XM_005253868.3 | XP_005253925.1 | 4, 5 | 612 | |||
| XM_017019175.1 | XP_016874664.1 | 5, 6 | 698 | |||
| XM_017019176.1 | XP_016874665.1 | 6, 7 | 930 | |||
| XM_017019177.1 | XP_016874666.1 | 6, 7 | 930 | |||
| GenBank mRNA | AK289621.1 | - | 5, 6 | 714 | ||
| BC037296.2 | - | 5, 6 | 921 | 4821240 | ||

FBXW8
Genomic Map