siRNA Details
| Gene Symbol: TAPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153365.2 | NP_699196.2 | 14 | 2908 | ||
| XM_005248139.2 | XP_005248196.1 | 13 | 2722 | |||
| XM_005248140.3 | XP_005248197.1 | 13 | 2672 | |||
| XM_011513812.2 | XP_011512114.1 | 13 | 2771 | |||
| XM_011513815.2 | XP_011512117.1 | 13 | 4007 | |||
| XM_011513816.2 | XP_011512118.1 | 13 | 2716 | |||
| XM_011513817.2 | XP_011512119.1 | 11 | 2285 | |||
| XM_017007876.1 | XP_016863365.1 | 15 | 2798 | |||
| GenBank mRNA | AK074494.1 | - | 15 | 2493 | ||

TAPT1
Genomic Map