siRNA Details
| Gene Symbol: PI16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199159.1 | NP_001186088.1 | 6 | 1335 | ||
| NM_153370.2 | NP_699201.2 | 5 | 1440 | |||
| XM_011514375.2 | XP_011512677.1 | 5 | 1269 | |||
| GenBank mRNA | AK075470.1 | - | 5 | 1135 | ||
| AK302193.1 | - | 4 | 943 | |||
| AK303928.1 | - | 4 | 1089 | |||
| AK312785.1 | - | 5 | 1133 | |||
| AY358422.1 | - | 5 | 1126 | |||
| BC022399.1 | - | 5 | 1134 | 4691115 | ||
| BC035634.1 | - | 5 | 1440 | 5217121 | ||

PI16
Genomic Map