siRNA Details
| Gene Symbol: COG7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153603.3 | NP_705831.1 | 1, 2 | 373 | ||
| GenBank mRNA | BC000549.2 | - | 1, 2 | 328 | 3163316 | |
| BC037563.1 | - | 1, 2 | 362 | 5393265 | ||
| BP308365.1 | - | 1, 2 | 364 | |||
| DB444997.1 | - | 1, 2 | 380 | |||
| KU178837.1 | - | 1, 2 | 176 | |||
| KU178838.1 | - | 1, 2 | 176 | |||

COG7
Genomic Map