siRNA Details
| Gene Symbol: PIANP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244014.1 | NP_001230943.1 | 3 | 729 | ||
| NM_001244015.1 | NP_001230944.1 | 3 | 795 | |||
| NM_153685.3 | NP_710152.1 | 3 | 729 | |||
| XM_011520926.2 | XP_011519228.1 | 3 | 686 | |||
| GenBank mRNA | AK056295.1 | - | 3 | 686 | ||
| AK091596.1 | - | 4 | 451 | |||
| AK091656.1 | - | 3 | 529 | |||
| AK095854.1 | - | 3 | 795 | |||
| AK289648.1 | - | 3 | 584 | |||
| AY358534.1 | - | 2 | 513 | |||
| BC035736.1 | - | 3 | 532 | 5730150 | ||

PIANP
Genomic Map