siRNA Details
| Gene Symbol: PGR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000926.4 | NP_000917.3 | 2 | 2477 | ||
| NM_001202474.3 | NP_001189403.1 | 2 | 1727 | |||
| NM_001271161.2 | NP_001258090.1 | 2 | 1727 | |||
| NM_001271162.1 | NP_001258091.1 | 2 | 397 | |||
| NR_073141.2 | 2 | 1727 | ||||
| NR_073142.2 | 2 | 1727 | ||||
| NR_073143.2 | 2 | 1727 | ||||
| XM_006718858.3 | XP_006718921.1 | 2 | 3327 | |||
| XM_011542869.2 | XP_011541171.1 | 2 | 3345 | |||
| GenBank mRNA | AB084248.1 | - | 2 | 1734 | ||
| AB085683.1 | - | 2 | 1734 | |||
| AB085843.1 | - | 2 | 1734 | |||
| AB085844.1 | - | 2 | 1734 | |||
| AB085845.1 | - | 2 | 1734 | |||
| AF016381.1 | - | 2 | 1909 | |||
| AK304853.1 | - | 2 | 397 | |||
| AY382151.1 | - | 1 | 93 | |||
| M15716.1 | - | 2 | 1909 | |||
| X51730.1 | - | 2 | 3188 | |||

PGR
Genomic Map