siRNA Details
| Gene Symbol: FNDC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171940.1 | NP_001165411.2 | 3 | 366 | ||
| NM_001171941.2 | NP_001165412.1 | 3 | 300 | |||
| NM_153756.2 | NP_715637.2 | 3 | 366 | |||
| GenBank mRNA | AK092102.1 | - | 3 | 299 | ||
| BC062297.1 | - | 3 | 289 | 6338770 | ||
| BE502835.1 | - | 3 | 367 | |||
| BX537781.1 | - | 3 | 242 | |||

FNDC5
Genomic Map