siRNA Details
| Gene Symbol: PHF13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153812.2 | NP_722519.2 | 2 | 447 | ||
| XM_011540762.1 | XP_011539064.1 | 2 | 249 | |||
| GenBank mRNA | AB209544.1 | - | 2 | 139 | ||
| AK097715.1 | - | 2 | 381 | |||
| AK315110.1 | - | 2 | 379 | |||
| AY069975.1 | - | 2 | 77 | |||
| BC038516.1 | - | 2 | 458 | 5266750 | ||
| DB064673.1 | - | 3 | 382 | |||

PHF13
Genomic Map