siRNA Details
| Gene Symbol: H1FOO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153833.2 | NP_722575.1 | 1 | 78 | ||
| GenBank mRNA | AY158091.1 | - | 1 | 78 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

H1FOO| Gene Symbol: H1FOO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153833.2 | NP_722575.1 | 1 | 78 | ||
| GenBank mRNA | AY158091.1 | - | 1 | 78 | ||