siRNA Details
| Gene Symbol: CSF3R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000760.3 | NP_000751.1 | 4 | 841 | ||
| NM_156039.3 | NP_724781.1 | 4 | 841 | |||
| NM_172313.2 | NP_758519.1 | 4 | 841 | |||
| XM_005270493.1 | XP_005270550.1 | 4 | 841 | |||
| XM_011540748.2 | XP_011539050.1 | 3 | 366 | |||
| XM_011540749.1 | XP_011539051.1 | 4 | 841 | |||
| XM_017000370.1 | XP_016855859.1 | 3 | 403 | |||
| GenBank mRNA | AB209656.1 | - | 1 | 847 | ||
| BC053585.1 | - | 4 | 434 | 5217305 | ||
| DQ424902.1 | - | 2 | 257 | |||
| M59818.1 | - | 4 | 426 | |||
| M59819.1 | - | 4 | 426 | |||
| M59820.1 | - | 4 | 426 | |||
| X55720.1 | - | 4 | 421 | |||
| X55721.1 | - | 4 | 422 | |||

CSF3R
Genomic Map