siRNA Details
| Gene Symbol: TAC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077503.1 | NP_001070971.1 | 1 | 80 | ||
| NM_001077504.1 | NP_001070972.1 | 1 | 80 | |||
| NM_001077505.1 | NP_001070973.1 | 1 | 80 | |||
| NM_001077506.1 | NP_001070974.1 | 1 | 80 | |||
| NM_170685.2 | NP_733786.2 | 1 | 80 | |||
| GenBank mRNA | AF515828.1 | - | 1 | 80 | ||
| AF515829.1 | - | 1 | 80 | |||
| AF515830.1 | - | 1 | 80 | |||
| AF515831.1 | - | 1 | 80 | |||
| AF521560.1 | - | 1 | 80 | |||
| AY471574.1 | - | 1 | 80 | |||
| BC100841.1 | - | 1 | 80 | |||

TAC4
Genomic Map