siRNA Details
Gene Symbol: HIST1H2AA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170745.3 | NP_734466.1 | 1 | 154 | ||
GenBank mRNA | BC062211.1 | - | 1 | 158 | 6614487 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HIST1H2AA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170745.3 | NP_734466.1 | 1 | 154 | ||
GenBank mRNA | BC062211.1 | - | 1 | 158 | 6614487 |