siRNA Details
| Gene Symbol: ADGRG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304376.2 | NP_001291305.1 | 6 | 617 | ||
| NM_001318481.1 | NP_001305410.1 | 6 | 617 | |||
| NM_153837.3 | NP_722579.1 | 6 | 1089 | |||
| XM_011522949.2 | XP_011521251.1 | 6 | 633 | |||
| XM_011522950.2 | XP_011521252.1 | 6 | 659 | |||
| GenBank mRNA | AK128298.1 | - | 6 | 768 | ||
| AK160368.1 | - | 6 | 599 | |||
| AK172765.1 | - | 2 | 106 | |||
| AK292783.1 | - | 6 | 609 | |||
| AK301357.1 | - | 6 | 537 | |||
| AK303661.1 | - | 6 | 609 | |||
| AY140956.1 | - | 5 | 514 | |||
| AY358447.1 | - | 6 | 596 | |||
| BC032401.1 | - | 6 | 1037 | 5210813 | ||
More Information
Set Membership: |
|

ADGRG5
Genomic Map