siRNA Details
| Gene Symbol: RAB39B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_171998.3 | NP_741995.1 | 1, 2 | 518 | ||
| GenBank mRNA | AL834460.1 | - | 1, 2 | 381 | ||
| AY052478.1 | - | 1, 2 | 416 | |||
| BC009714.1 | - | 1, 2 | 518 | 3900092 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAB39B| Gene Symbol: RAB39B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_171998.3 | NP_741995.1 | 1, 2 | 518 | ||
| GenBank mRNA | AL834460.1 | - | 1, 2 | 381 | ||
| AY052478.1 | - | 1, 2 | 416 | |||
| BC009714.1 | - | 1, 2 | 518 | 3900092 | ||