siRNA Details
| Gene Symbol: HSCB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318316.1 | NP_001305245.1 | 4 | 281 | ||
| NM_172002.4 | NP_741999.3 | 4 | 527 | |||
| NR_134560.1 | 4 | 472 | ||||
| XM_005261359.3 | XP_005261416.1 | 4 | 528 | |||
| XM_017028619.1 | XP_016884108.1 | 4 | 498 | |||
| XM_017028620.1 | XP_016884109.1 | 4 | 331 | |||
| GenBank mRNA | AY191719.1 | - | 4 | 477 | ||
| BC000004.1 | - | 4 | 536 | |||
| BC065569.1 | - | 4 | 500 | 6171959 | ||
| CR456462.1 | - | 4 | 495 | |||
| DW414316.1 | - | 4 | 292 | |||
| HY057929.1 | - | 4 | 281 | |||

HSCB
Genomic Map