siRNA Details
| Gene Symbol: WFDC13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172005.1 | NP_742002.1 | 3 | 368 | ||
| GenBank mRNA | AF454505.1 | - | 3 | 368 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

WFDC13| Gene Symbol: WFDC13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172005.1 | NP_742002.1 | 3 | 368 | ||
| GenBank mRNA | AF454505.1 | - | 3 | 368 | ||