siRNA Details
| Gene Symbol: RTP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153708.2 | NP_714919.2 | 2 | 1496 | ||
| GenBank mRNA | BC034744.1 | - | 2 | 1457 | ||
| BC065202.1 | - | 2 | 1497 | 5179096 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RTP1| Gene Symbol: RTP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153708.2 | NP_714919.2 | 2 | 1496 | ||
| GenBank mRNA | BC034744.1 | - | 2 | 1457 | ||
| BC065202.1 | - | 2 | 1497 | 5179096 | ||