siRNA Details
| Gene Symbol: ADGRG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304376.2 | NP_001291305.1 | 12 | 2117 | ||
| NM_001318481.1 | NP_001305410.1 | 11 | 1999 | |||
| NM_153837.3 | NP_722579.1 | 12 | 2589 | |||
| XM_011522949.2 | XP_011521251.1 | 12 | 2133 | |||
| XM_011522950.2 | XP_011521252.1 | 12 | 2159 | |||
| GenBank mRNA | AK128298.1 | - | 12 | 2268 | ||
| AK160368.1 | - | 11 | 3547 | |||
| AK172765.1 | - | 8 | 1606 | |||
| AK292783.1 | - | 12 | 2109 | |||
| AY358447.1 | - | 12 | 2096 | |||
| BC032401.1 | - | 12 | 2537 | 5210813 | ||
More Information
Set Membership: |
|

ADGRG5
Genomic Map