siRNA Details
| Gene Symbol: WEE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143976.1 | NP_001137448.1 | 2 | 789 | ||
| NM_003390.3 | NP_003381.1 | 2 | 929 | |||
| GenBank mRNA | AK122837.1 | - | 2 | 789 | ||
| BC051831.1 | - | 2 | 727 | 6147695 | ||
| BC070052.1 | - | 2 | 919 | 30336707 | ||
| BX641032.1 | - | 1 | 447 | |||
| U10564.1 | - | 2 | 929 | |||
| X62048.1 | - | 2 | 846 | |||

WEE1
Genomic Map