siRNA Details
| Gene Symbol: WEE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143976.1 | NP_001137448.1 | 6 | 1365 | ||
| NM_003390.3 | NP_003381.1 | 6 | 1505 | |||
| GenBank mRNA | AK122837.1 | - | 6 | 1365 | ||
| BC051831.1 | - | 6 | 1303 | 6147695 | ||
| BC070052.1 | - | 6 | 1495 | 30336707 | ||
| U10564.1 | - | 6 | 1505 | |||
| X62048.1 | - | 6 | 1422 | |||

WEE1
Genomic Map