siRNA Details
| Gene Symbol: MRPL54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172251.2 | NP_758455.1 | 3 | 515 | ||
| GenBank mRNA | BC065273.1 | - | 3 | 508 | 6165136 | |
| BF346319.1 | - | 3 | 508 | |||
| BM792364.1 | - | 3 | 475 | |||
| BQ061807.1 | - | 3 | 503 | |||

MRPL54
Genomic Map